2-(4-Fluorophenoxy)-1-methyl-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044173-500MG 044173-1G 044173-5G
101864-534EA 747.69 USD
101864-534 101864-536 101864-538
2-(4-Fluorophenoxy)-1-methyl-1H-indole-3-carbaldehyde ≥95%
2-(4-Fluorophenoxy)-1-methyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₂FNO₂
Melting Pt: 139…141 °C
MDL Number: MFCD01315828
CAS Number: 338416-70-9

Order Now

Specification Test Results

Melting point 139-141°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR