2-(4-Chlorophenoxy)-1-methyl-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044171-500MG 044171-1G 044171-5G
101864-522EA 747.69 USD
101864-522 101864-524 101864-526
2-(4-Chlorophenoxy)-1-methyl-1H-indole-3-carbaldehyde ≥95%
2-(4-Chlorophenoxy)-1-methyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₂ClNO₂
Melting Pt: 156…158 °C
MDL Number: MFCD01315827
CAS Number: 338416-66-3

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR