2-[(2-Chlorophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde ≥95%
Supplier: Matrix Scientific
044169-500MG
044169-1G
044169-5G
101864-510EA
747.69
USD
101864-510
101864-512
101864-514
2-[(2-Chlorophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde ≥95%
2-[(2-Chlorophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde
Hazard: Irritant
Formula:
C₁₆H₁₂ClNOS Melting Pt: 162…164 °C |
MDL Number:
MFCD01315818 CAS Number: 338416-38-9 |
Specification Test Results
Melting point | 162-164°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...