2-[(2,6-Dichlorophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044168-500MG 044168-1G 044168-5G
101864-504EA 747.69 USD
101864-504 101864-506 101864-508
2-[(2,6-Dichlorophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde ≥95%
2-[(2,6-Dichlorophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₁Cl₂NOS
Melting Pt: 186…190 °C
MDL Number: MFCD01568345
CAS Number: 338416-35-6

Order Now

Specification Test Results

Melting point 186-190°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR