1-Methyl-2-[(4-methylphenyl)sulfanyl]-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044166-500MG 044166-1G 044166-5G
101864-492EA 747.69 USD
101864-492 101864-494 101864-496
1-Methyl-2-[(4-methylphenyl)sulfanyl]-1H-indole-3-carbaldehyde ≥95%
1-Methyl-2-[(4-methylphenyl)sulfanyl]-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₇H₁₅NOS
Melting Pt: 132…133 °C
MDL Number: MFCD01315812
CAS Number: 338416-19-6

Order Now

Specification Test Results

Melting point 132-133°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR