2-[(4-Bromophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

044164-500MG 044164-1G 044164-5G
101864-480EA 747.69 USD
101864-480 101864-482 101864-484
2-[(4-Bromophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde ≥95%
2-[(4-Bromophenyl)sulfanyl]-1-methyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₂BrNOS
Melting Pt: 156…158 °C
MDL Number: MFCD01315810
CAS Number: 338416-15-2

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR