5-Acetyl-6-[2-(dimethylamino)vinyl]-2(1H)-pyridinone ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Acetyl-6-[(E)-2-(dimethylamino)ethenyl]-1,2-dihydropyridin-2-one
044162-500MG
044162-1G
101864-472EA
1109.23
USD
101864-472
101864-474
5-Acetyl-6-[2-(dimethylamino)vinyl]-2(1H)-pyridinone ≥95%
5-Acetyl-6-[2-(dimethylamino)vinyl]-2(1H)-pyridinone
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₂ MW: 206.25 g/mol Melting Pt: 238…240 °C |
MDL Number:
MFCD01814813 CAS Number: 88877-00-3 |
Specification Test Results
Melting point | 238-240°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...