5-Acetyl-6-methyl-2(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Acetyl-6-methyl-1,2-dihydropyridin-2-one, 5-Acetyl-6-methylpyridin-2(1H)-one

044161-500MG 044161-1G
101864-468EA 1000 USD
101864-468 101864-470
5-Acetyl-6-methyl-2(1H)-pyridinone ≥95%
5-Acetyl-6-methyl-2(1H)-pyridinone

Hazard: Irritant

Formula: C₈H₉NO₂
MW: 151.17 g/mol
Melting Pt: 203…205 °C
MDL Number: MFCD01814812
CAS Number: 5220-65-5

Order Now

Specification Test Results

Melting point 203-205°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR