8-Amino-3-(4-chlorophenyl)imidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Chlorophenyl)imidazo[1,2-a]pyridin-8-amine

044157-500MG 044157-1G 044157-5G
101864-444EA 747.69 USD
101864-444 101864-446 101864-448
8-Amino-3-(4-chlorophenyl)imidazo[1,2-a]pyridine ≥95%
8-Amino-3-(4-chlorophenyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₁₃H₁₀ClN₃
Melting Pt: 151-161°C
MDL Number: MFCD00975015
CAS Number: 338415-10-4

Order Now

Specification Test Results

Melting point 151-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR