Methyl-3-{[(benzoylamino)carbothioyl]amino}-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

044155-500MG 044155-1G 044155-5G
101864-432EA 278.46 USD
101864-432 101864-434 101864-436
Methyl-3-{[(benzoylamino)carbothioyl]amino}-2-thiophenecarboxylate ≥95%
Methyl-3-{[(benzoylamino)carbothioyl]amino}-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₃S₂
Melting Pt: 144…146 °C
MDL Number: MFCD00955295
CAS Number: 178675-17-7

Order Now

Specification Test Results

Melting point 144-146°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR