6-(Methoxymethyl)-2-methyl-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

044153-500MG 044153-1G 044153-5G
101864-426EA 181.54 USD
101864-426 101864-428 101864-430
6-(Methoxymethyl)-2-methyl-4-pyrimidinol ≥95%
6-(Methoxymethyl)-2-methyl-4-pyrimidinol

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
Melting Pt: 178…181 °C
MDL Number: MFCD00087858
CAS Number: 38249-50-2

Order Now

Specification Test Results

Melting point 178-181°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR