1-[2-(4-Chlorophenyl)-1,3-thiazol-5-yl]-3-(dimethylamino)-2-propen-1-one ≥95%
Supplier: Matrix Scientific
044150-500MG
044150-1G
101864-416EA
747.69
USD
101864-416
101864-418
1-[2-(4-Chlorophenyl)-1,3-thiazol-5-yl]-3-(dimethylamino)-2-propen-1-one ≥95%
1-[2-(4-Chlorophenyl)-1,3-thiazol-5-yl]-3-(dimethylamino)-2-propen-1-one
Hazard: Irritant
Formula:
C₁₄H₁₃ClN₂OS Melting Pt: 233…235 °C |
MDL Number:
MFCD00974303 CAS Number: 338414-64-5 |
Specification Test Results
Melting point | 233-235°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...