2-Amino-6-(6-fluoro-2-pyridinyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

044147-500MG 044147-1G 044147-5G
101864-398EA 472.31 USD
101864-398 101864-400 101864-402
2-Amino-6-(6-fluoro-2-pyridinyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile ≥95%
2-Amino-6-(6-fluoro-2-pyridinyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile

Hazard: Irritant

Formula: C₁₃H₁₁FN₄S
MW: 274.32 g/mol
Melting Pt: 209…211 °C
MDL Number: MFCD00794643
CAS Number: 338413-86-8

Order Now

Specification Test Results

Melting point 209-211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR