1-(6-Chloro-3-pyridazinyl)-4-piperidinecarbohydrazide ≥95%

Supplier: Matrix Scientific

044145-500MG 044145-1G 044145-5G
101864-386EA 1109.23 USD
101864-386 101864-388 101864-390
1-(6-Chloro-3-pyridazinyl)-4-piperidinecarbohydrazide ≥95%
1-(6-Chloro-3-pyridazinyl)-4-piperidinecarbohydrazide

Hazard: Irritant

Formula: C₁₀H₁₄ClN₅O
Melting Pt: 209-211°C
MDL Number: MFCD00794640
CAS Number: 338413-80-2

Order Now

Specification Test Results

Melting point 209-211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR