(5-Chloro-1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazol-4-yl)methanol ≥95%

Supplier: Matrix Scientific

044143-500MG 044143-1G 044143-5G
101864-374EA 921.54 USD
101864-374 101864-376 101864-378
(5-Chloro-1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazol-4-yl)methanol ≥95%
(5-Chloro-1-methyl-3-{[(4-methylphenyl)sulfanyl]methyl}-1H-pyrazol-4-yl)methanol

Hazard: Irritant

Formula: C₁₃H₁₅ClN₂OS
Melting Pt: 103…105 °C
MDL Number: MFCD00794886
CAS Number: 318234-43-4

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR