(5-Chloro-3-{[(4-chlorophenyl)sulfanyl]methyl}-1-methyl-1H-pyrazol-4-yl)methanol ≥95%

Supplier: Matrix Scientific

044142-500MG 044142-1G 044142-5G
101864-368EA 921.54 USD
101864-368 101864-370 101864-372
(5-Chloro-3-{[(4-chlorophenyl)sulfanyl]methyl}-1-methyl-1H-pyrazol-4-yl)methanol ≥95%
(5-Chloro-3-{[(4-chlorophenyl)sulfanyl]methyl}-1-methyl-1H-pyrazol-4-yl)methanol

Hazard: Irritant

Formula: C₁₂H₁₂Cl₂N₂OS
Melting Pt: 104…105 °C
MDL Number: MFCD00794871
CAS Number: 318234-28-5

Order Now

Specification Test Results

Melting point 104-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR