5-Chloro-3-{[(4-chlorophenyl)sulfanyl]methyl}-1-methyl-1H-pyrazole-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
044141-500MG
044141-1G
044141-5G
101864-362EA
747.69
USD
101864-362
101864-364
101864-366
5-Chloro-3-{[(4-chlorophenyl)sulfanyl]methyl}-1-methyl-1H-pyrazole-4-carbaldehyde ≥95%
5-Chloro-3-{[(4-chlorophenyl)sulfanyl]methyl}-1-methyl-1H-pyrazole-4-carbaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₀Cl₂N₂OS Melting Pt: 105…108 °C |
MDL Number:
MFCD00794870 CAS Number: 318234-27-4 |
Specification Test Results
Melting point | 105-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...