5-Chloro-1-methyl-3-[(phenylsulfanyl)methyl]-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-1-methyl-3-[(phenylthio)methyl]-1H-pyrazole-4-carboxaldehyde

044140-500MG 044140-1G 044140-5G
101864-356EA 510.77 USD
101864-356 101864-358 101864-360
5-Chloro-1-methyl-3-[(phenylsulfanyl)methyl]-1H-pyrazole-4-carbaldehyde ≥95%
5-Chloro-1-methyl-3-[(phenylsulfanyl)methyl]-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂OS
MW: 266.75 g/mol
Melting Pt: 105…107 °C
MDL Number: MFCD00794869
CAS Number: 318234-26-3

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR