{2-[(4-Chlorophenyl)sulfanyl]-3-pyridinyl}methanol ≥95%

Supplier: Matrix Scientific

044139-500MG 044139-1G 044139-5G
101864-350EA 556.92 USD
101864-350 101864-352 101864-354
{2-[(4-Chlorophenyl)sulfanyl]-3-pyridinyl}methanol ≥95%
{2-[(4-Chlorophenyl)sulfanyl]-3-pyridinyl}methanol

Hazard: Irritant

Formula: C₁₂H₁₀ClNOS
MW: 251.74 g/mol
Melting Pt: 93…95 °C
MDL Number: MFCD00794373
CAS Number: 51723-83-2

Order Now

Specification Test Results

Melting point 93-95°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR