{2-[(2-Chlorophenyl)sulfanyl]-3-pyridinyl}methanol ≥95%

Supplier: Matrix Scientific

044138-500MG 044138-1G 044138-5G
101864-344EA 556.92 USD
101864-344 101864-346 101864-348
{2-[(2-Chlorophenyl)sulfanyl]-3-pyridinyl}methanol ≥95%
{2-[(2-Chlorophenyl)sulfanyl]-3-pyridinyl}methanol

Hazard: Irritant

Formula: C₁₂H₁₀ClNOS
Melting Pt: 92…94 °C
MDL Number: MFCD00794372
CAS Number: 338982-34-6

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR