{2-[(4-Bromophenyl)sulfanyl]-3-pyridinyl}methanol ≥95%

Supplier: Matrix Scientific

044137-500MG 044137-1G 044137-5G
101864-338EA 556.92 USD
101864-338 101864-340 101864-342
{2-[(4-Bromophenyl)sulfanyl]-3-pyridinyl}methanol ≥95%
{2-[(4-Bromophenyl)sulfanyl]-3-pyridinyl}methanol

Hazard: Irritant

Formula: C₁₂H₁₀BrNOS
Melting Pt: 94…95.5 °C
MDL Number: MFCD00794361
CAS Number: 338982-33-5

Order Now

Specification Test Results

Melting point 94-95.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR