2-(4-Chlorophenoxy)nicotinaldehyde ≥95%
Supplier: Matrix Scientific
044136-500MG
044136-1G
044136-5G
101864-332EA
375.38
USD
101864-332
101864-334
101864-336
2-(4-Chlorophenoxy)nicotinaldehyde ≥95%
2-(4-Chlorophenoxy)nicotinaldehyde
Hazard: Irritant
Formula:
C₁₂H₈ClNO₂ Melting Pt: 90…92 °C |
MDL Number:
MFCD00794360 CAS Number: 478031-03-7 |
Specification Test Results
Melting point | 90-92°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...