2-(4-Methylphenoxy)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Methylphenoxy)pyridine-3-carboxaldehyde

044135-500MG 044135-1G 044135-5G
101864-326EA 246.15 USD
101864-326 101864-328 101864-330
2-(4-Methylphenoxy)nicotinaldehyde ≥95%
2-(4-Methylphenoxy)nicotinaldehyde

Hazard: Irritant

Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
Melting Pt: 77…80 °C
MDL Number: MFCD00794346
CAS Number: 478030-89-6

Order Now

Specification Test Results

Melting point 77-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR