2-[(2-Chlorophenyl)sulfanyl]nicotinaldehyde ≥95%

Supplier: Matrix Scientific

044134-500MG 044134-1G 044134-5G
101864-320EA 247.69 USD
101864-320 101864-322 101864-324
2-[(2-Chlorophenyl)sulfanyl]nicotinaldehyde ≥95%
2-[(2-Chlorophenyl)sulfanyl]nicotinaldehyde

Hazard: Irritant

Formula: C₁₂H₈ClNOS
MW: 249.72 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD00794345
CAS Number: 338982-32-4

Order Now

Specification Test Results

Melting point 108-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR