2-(Phenylsulfanyl)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

044133-500MG 044133-1G 044133-5G
101864-314EA 376.92 USD
101864-314 101864-316 101864-318
2-(Phenylsulfanyl)nicotinaldehyde ≥95%
2-(Phenylsulfanyl)nicotinaldehyde

Hazard: Irritant

Formula: C₁₂H₉NOS
MW: 215.28 g/mol
Melting Pt: 81.5…84 °C
MDL Number: MFCD00794344
CAS Number: 338982-31-3

Order Now

Specification Test Results

Melting point 81.5-84°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR