2-[(4-Bromophenyl)sulfanyl]nicotinaldehyde ≥95%

Supplier: Matrix Scientific

044132-500MG 044132-1G 044132-5G
101864-308EA 747.69 USD
101864-308 101864-310 101864-312
2-[(4-Bromophenyl)sulfanyl]nicotinaldehyde ≥95%
2-[(4-Bromophenyl)sulfanyl]nicotinaldehyde

Hazard: Irritant

Formula: C₁₂H₈BrNOS
Melting Pt: 101…103 °C
MDL Number: MFCD00794343
CAS Number: 338982-30-2

Order Now

Specification Test Results

Melting point 101-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR