2-[(2,4-Dichlorophenyl)sulfanyl]nicotinaldehyde ≥95%

Supplier: Matrix Scientific

044131-500MG 044131-1G 044131-5G
101864-302EA 747.69 USD
101864-302 101864-304 101864-306
2-[(2,4-Dichlorophenyl)sulfanyl]nicotinaldehyde ≥95%
2-[(2,4-Dichlorophenyl)sulfanyl]nicotinaldehyde

Hazard: Irritant

Formula: C₁₂H₇Cl₂NOS
MW: 284.17 g/mol
Melting Pt: 114…117 °C
MDL Number: MFCD00794342
CAS Number: 338982-29-9

Order Now

Specification Test Results

Melting point 114-117°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR