2-[(4-Chlorophenyl)sulfanyl]nicotinaldehyde ≥95%

Supplier: Matrix Scientific

044130-500MG 044130-1G 044130-5G
101864-296EA 376.92 USD
101864-296 101864-298 101864-300
2-[(4-Chlorophenyl)sulfanyl]nicotinaldehyde ≥95%
2-[(4-Chlorophenyl)sulfanyl]nicotinaldehyde

Hazard: Irritant

Formula: C₁₂H₈ClNOS
MW: 249.72 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD00794341
CAS Number: 338982-28-8

Order Now

Specification Test Results

Melting point 86-88°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR