3-Chloro-4-(2-pyrimidinyloxy)phenylamine ≥95%

Supplier: Matrix Scientific

044128-500MG 044128-1G
101864-286EA 307.69 USD
101864-286 101864-288
3-Chloro-4-(2-pyrimidinyloxy)phenylamine ≥95%
3-Chloro-4-(2-pyrimidinyloxy)phenylamine

Hazard: Irritant

Formula: C₁₀H₈ClN₃O
Melting Pt: 173…175 °C
MDL Number: MFCD00244331
CAS Number: 338413-13-1

Order Now

Specification Test Results

Melting point 173-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR