5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

044125-500MG 044125-1G 044125-5G
101864-268EA 747.69 USD
101864-268 101864-270 101864-272
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylic acid ≥95%
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₂
Melting Pt: 176…177 °C
MDL Number: MFCD01314939
CAS Number: 191419-14-4

Order Now

Specification Test Results

Melting point 176-177°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR