2-Chloro-4,6-dimethyl-3-(methylsulfonyl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3-methanesulphonyl-4,6-dimethylpyridine

044122-500MG 044122-1G
101864-256EA 556.92 USD
101864-256 101864-258
2-Chloro-4,6-dimethyl-3-(methylsulfonyl)pyridine ≥95%
2-Chloro-4,6-dimethyl-3-(methylsulfonyl)pyridine

Hazard: Irritant

Formula: C₈H₁₀ClNO₂S
MW: 219.69 g/mol
Melting Pt: 131…133 °C
MDL Number: MFCD08056620
CAS Number: 886361-59-7

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR