Dimethyl-1,3-thiazolane-2,4-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Thiazolidine-2,4-dicarboxylic acid dimethyl ester

044121-500MG 044121-5G
101864-250EA 747.69 USD
101864-250 101864-254
Dimethyl-1,3-thiazolane-2,4-dicarboxylate ≥95%
Dimethyl-1,3-thiazolane-2,4-dicarboxylate

Hazard: Irritant

Formula: C₇H₁₁NO₄S
MW: 205.23 g/mol
Melting Pt: 72…74 °C
MDL Number: MFCD00191687
CAS Number: 318233-97-5

Order Now

Specification Test Results

Melting point 72-74°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR