Thiazolidine-2,4-dicarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Tidiacic, Tetrahydrothiazole-2,4-dicarboxylic acid

044120-500MG 044120-1G 044120-5G
101864-244EA 52.31 USD
101864-244 101864-246 101864-248
Thiazolidine-2,4-dicarboxylic acid ≥95%
Thiazolidine-2,4-dicarboxylic acid

Hazard: Irritant

Formula: C₅H₇NO₄S
MW: 177.1784 g/mol
Melting Pt: 199-201°C
MDL Number: MFCD00145399
CAS Number: 30097-06-4

Order Now

Specification Test Results

Melting point 199-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR