[3-(Bromomethyl)-1-benzofuran-2-yl](phenyl)methanone ≥95%

Supplier: Matrix Scientific

044116-500MG 044116-1G 044116-5G
101864-226EA 747.69 USD
101864-226 101864-228 101864-230
[3-(Bromomethyl)-1-benzofuran-2-yl](phenyl)methanone ≥95%
[3-(Bromomethyl)-1-benzofuran-2-yl](phenyl)methanone

Hazard: Irritant

Formula: C₁₆H₁₁BrO₂
Melting Pt: 104…106 °C
MDL Number: MFCD05663788
CAS Number: 67534-81-0

Order Now

Specification Test Results

Melting point 104-106°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR