(Z)-2-(4-Chlorophenyl)-3-(dimethylamino)-2-propenal ≥95%

Supplier: Matrix Scientific

044115-500MG 044115-1G 044115-5G
101864-220EA 278.46 USD
101864-220 101864-222 101864-224
(Z)-2-(4-Chlorophenyl)-3-(dimethylamino)-2-propenal ≥95%
(Z)-2-(4-Chlorophenyl)-3-(dimethylamino)-2-propenal

Hazard: Irritant

Formula: C₁₁H₁₂ClNO
Melting Pt: 123…125 °C
MDL Number: MFCD04973720
CAS Number: 85907-66-0

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR