5-Methoxy-2-sulfanyl-4-pyrimidinol ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Methoxy-2-thioxo-2,3-dihydropyrimidin-4(1H)-one
044111-500MG
044111-1G
044111-5G
101864-200EA
375.38
USD
101864-200
101864-202
101864-204
5-Methoxy-2-sulfanyl-4-pyrimidinol ≥95%
5-Methoxy-2-sulfanyl-4-pyrimidinol
Hazard: Irritant
Formula:
C₅H₆N₂O₂S MW: 158.18 g/mol Melting Pt: 280…281 °C |
MDL Number:
MFCD00094470 CAS Number: 6939-11-3 |
Specification Test Results
Melting point | 255-266°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...