1-(4-Fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
044109-500MG
044109-1G
044109-5G
101864-192EA
375.38
USD
101864-192
101864-194
101864-196
1-(4-Fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylic acid ≥95%
1-(4-Fluorobenzoyl)-4-hydroxy-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₂FNO₄ MW: 253.23 g/mol Melting Pt: 205…208 °C |
MDL Number:
MFCD00202775 CAS Number: 497163-88-9 |
Specification Test Results
Melting point | 205-208°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...