3-{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-indol-3-yl}acrylic acid ≥95%

Supplier: Matrix Scientific

044108-500MG 044108-1G 044108-5G
101864-186EA 1109.23 USD
101864-186 101864-188 101864-190
3-{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-indol-3-yl}acrylic acid ≥95%
3-{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-indol-3-yl}acrylic acid

Hazard: Irritant

Formula: C₁₇H₁₀ClF₃N₂O₂
Melting Pt: 250…253 °C
MDL Number: MFCD00664519
CAS Number: 1164457-90-2

Order Now

Specification Test Results

Melting point 250-253°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR