Methyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}butanoate ≥95%

Supplier: Matrix Scientific

044107-500MG 044107-1G 044107-5G
101864-180EA 747.69 USD
101864-180 101864-182 101864-184
Methyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}butanoate ≥95%
Methyl-4-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]amino}butanoate

Hazard: Irritant

Formula: C₁₁H₁₂ClF₃N₂O₂
MW: 296.68 g/mol
Melting Pt: 46…48 °C
MDL Number: MFCD00172609
CAS Number: 332361-10-1

Order Now

Specification Test Results

Melting point 46-48°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR