2-{[2-(3,4-Dimethoxyanilino)-2-oxoethyl]sulfanyl}acetic acid ≥95%

Supplier: Matrix Scientific

044106-500MG 044106-1G 044106-5G
101864-174EA 181.54 USD
101864-174 101864-176 101864-178
2-{[2-(3,4-Dimethoxyanilino)-2-oxoethyl]sulfanyl}acetic acid ≥95%
2-{[2-(3,4-Dimethoxyanilino)-2-oxoethyl]sulfanyl}acetic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₅S
Melting Pt: 128…130 °C
MDL Number: MFCD00202285
CAS Number: 338409-77-1

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR