2-{[2-(2,4-Dichloroanilino)-2-oxoethyl]sulfanyl}acetic acid ≥95%
Supplier: Matrix Scientific
044105-500MG
044105-1G
044105-5G
101864-168EA
181.54
USD
101864-168
101864-170
101864-172
2-{[2-(2,4-Dichloroanilino)-2-oxoethyl]sulfanyl}acetic acid ≥95%
2-{[2-(2,4-Dichloroanilino)-2-oxoethyl]sulfanyl}acetic acid
Hazard: Irritant
Formula:
C₁₀H₉Cl₂NO₃S Melting Pt: 125…126 °C |
MDL Number:
MFCD00202284 CAS Number: 338409-76-0 |
Specification Test Results
Melting point | 125-126°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...