2-{[2-(3,4-Dichloroanilino)-2-oxoethyl]sulfanyl}acetic acid ≥95%

Supplier: Matrix Scientific

044104-500MG 044104-1G 044104-5G
101864-162EA 181.54 USD
101864-162 101864-164 101864-166
2-{[2-(3,4-Dichloroanilino)-2-oxoethyl]sulfanyl}acetic acid ≥95%
2-{[2-(3,4-Dichloroanilino)-2-oxoethyl]sulfanyl}acetic acid

Hazard: Irritant

Formula: C₁₀H₉Cl₂NO₃S
Melting Pt: 181…183 °C
MDL Number: MFCD00202283
CAS Number: 70648-81-6

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR