2-[(2-Anilino-2-oxoethyl)sulfanyl]acetic acid ≥95%
Supplier: Matrix Scientific
044102-500MG
044102-1G
044102-5G
101864-150EA
181.54
USD
101864-150
101864-152
101864-154
2-[(2-Anilino-2-oxoethyl)sulfanyl]acetic acid ≥95%
2-[(2-Anilino-2-oxoethyl)sulfanyl]acetic acid
Hazard: Irritant
Formula:
C₁₀H₁₁NO₃S Melting Pt: 106…108 °C |
MDL Number:
MFCD00202279 CAS Number: 70648-87-2 |
Specification Test Results
Melting point | 106-108°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...