4-(4-Chlorophenyl)-1,2,3-thiadiazole-5-thiol ≥95%

Supplier: Matrix Scientific

044101-500MG 044101-1G 044101-5G
101864-144EA 747.69 USD
101864-144 101864-146 101864-148
4-(4-Chlorophenyl)-1,2,3-thiadiazole-5-thiol ≥95%
4-(4-Chlorophenyl)-1,2,3-thiadiazole-5-thiol

Hazard: Irritant

Formula: C₈H₅ClN₂S₂
Melting Pt: 108…110 °C
MDL Number: MFCD00173589
CAS Number: 338408-98-3

Order Now

Specification Test Results

Melting point 108-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR