4-Phenyl-1,2,3-thiadiazol-5-ylhydrosulfide ≥95%

Supplier: Matrix Scientific

044099-500MG 044099-1G 044099-5G
101864-132EA 747.69 USD
101864-132 101864-134 101864-136
4-Phenyl-1,2,3-thiadiazol-5-ylhydrosulfide ≥95%
4-Phenyl-1,2,3-thiadiazol-5-ylhydrosulfide

Hazard: Irritant

Formula: C₈H₆N₂S₂
Melting Pt: 80…82 °C
MDL Number: MFCD00173577
CAS Number: 215296-45-0

Order Now

Specification Test Results

Melting point 80-82°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR