4-(2-Bromoethoxy)benzaldehyde ≥95%

Supplier: Matrix Scientific

044093-500MG 044093-1G 044093-5G
101864-096EA 169.23 USD
101864-096 101864-098 101864-100
4-(2-Bromoethoxy)benzaldehyde ≥95%
4-(2-Bromoethoxy)benzaldehyde

Hazard: Irritant

Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
Melting Pt: 61…64 °C
MDL Number: MFCD00021074
CAS Number: 52191-15-8

Order Now

Specification Test Results

Melting point 61-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR