Ethyl-5-methyl-4-[(1,3-thiazol-2-ylamino)carbonyl]-3-isoxazolecarboxylate ≥95%
Supplier: Matrix Scientific
044089-500MG
044089-1G
101864-076EA
1109.23
USD
101864-076
101864-078
Ethyl-5-methyl-4-[(1,3-thiazol-2-ylamino)carbonyl]-3-isoxazolecarboxylate ≥95%
Ethyl-5-methyl-4-[(1,3-thiazol-2-ylamino)carbonyl]-3-isoxazolecarboxylate
Hazard: Irritant
Formula:
C₁₁H₁₁N₃O₄S Melting Pt: 154…156 °C |
MDL Number:
MFCD00664753 CAS Number: 338408-87-0 |
Specification Test Results
Melting point | 154-156°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...