Dimethyl-2-(hydroxyimino)malonate ≥95%

Supplier: Matrix Scientific

044088-500MG 044088-1G 044088-5G
101864-070EA 181.54 USD
101864-070 101864-072 101864-074
Dimethyl-2-(hydroxyimino)malonate ≥95%
Dimethyl-2-(hydroxyimino)malonate

Hazard: Irritant

Formula: C₅H₇NO₅
Melting Pt: 70…72 °C
MDL Number: MFCD00136215
CAS Number: 42937-74-6

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR