Ethyl-4-(4-bromobenzoyl)-3-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

044087-500MG 044087-1G
101864-066EA 1109.23 USD
101864-066 101864-068
Ethyl-4-(4-bromobenzoyl)-3-isoxazolecarboxylate ≥95%
Ethyl-4-(4-bromobenzoyl)-3-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₀BrNO₄
Melting Pt: 78…80 °C
MDL Number: MFCD00664751
CAS Number: 338408-83-6

Order Now

Specification Test Results

Melting point 78-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR