4-Methyl-2-(2-thienyl)-5-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-2-(thiophen-2-yl)thiazole-5-carboxylic acid

044085-500MG 044085-1G 044085-5G
101864-054EA 118.46 USD
101864-054 101864-056 101864-058
4-Methyl-2-(2-thienyl)-5-thiazolecarboxylic acid ≥95%
4-Methyl-2-(2-thienyl)-5-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₉H₇NO₂S₂
MW: 225.290 g/mol
Melting Pt: 233-235°C
MDL Number: MFCD01936004
CAS Number: 209540-08-9

Order Now

Specification Test Results

Melting point 233-235°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR